Order Entry
Finland
ContactUsLinkComponent
7-Amino-2-methyl-2H-1,4-benzoxazin-3(4H)-one
Irritant
7-Amino-2-methyl-2H-1,4-benzoxazin-3(4H)-one
Catalog # APOSOR40372-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
7-Amino-2-methyl-2H-1,4-benzoxazin-3(4H)-one
Catalog # APOSOR40372-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :178.188 g/mol
Melting Pt:173…175 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:105807-79-2
Formula:C₉H₁₀N₂O₂
MDL Number:MFCD09882585