

Methyl 2-(bromomethyl)-3-iodobenzoate 93%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:312747-81-2
:
Formula:C₉H₈BrIO₂
MDL Number:MFCD27949192
:354.967 g/mol
UN:3261
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Refrigerator