Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Bromomethyl)phenyl acetate
4-(Bromomethyl)phenyl acetate 97%, tech
4-(Bromomethyl)phenyl acetate 97%, tech
Corrosive
4-(Bromomethyl)phenyl acetate 97%, tech
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
4-(Bromomethyl)phenyl acetate 97%, tech
Supplier:
Apollo Scientific
About this item
CAS Number:
52727-95-4
:
Formula:
C₉H₉BrO₂
MDL Number:
MFCD17013576
:
229.071 g/mol
Storage Temperature:
Freezer
UN:
3261
ADR:
8,III
:
GHS05