Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Iodo-2-nitrophenol
5-Iodo-2-nitrophenol
5-Iodo-2-nitrophenol
5-Iodo-2-nitrophenol
Supplier:
Apollo Scientific
Certificates
Expand image
5-Iodo-2-nitrophenol
Supplier:
Apollo Scientific
About this item
CAS Number:
27783-55-7
:
Formula:
C₆H₄INO₃
MDL Number:
MFCD12923199
:
265.005 g/mol
Melting Pt:
92…96 °C
:
GHS07
Storage Temperature:
Ambient