Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-methoxy-2-naphthoate
Methyl 4-methoxy-2-naphthoate ≥98%
Methyl 4-methoxy-2-naphthoate ≥98%
Methyl 4-methoxy-2-naphthoate ≥98%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl 4-methoxy-2-naphthoate ≥98%
Supplier:
Apollo Scientific
About this item
CAS Number:
33295-47-5
:
Formula:
C₁₃H₁₂O₃
MDL Number:
MFCD13181453
:
216.233 g/mol
Storage Temperature:
Ambient