2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzonitrile, tech
Catalog # APOSOR33403-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.248 g/mol
Storage Temperature:Ambient
CAS Number:124678-40-6
Formula:C₁₃H₁₂N₂
MDL Number:MFCD01964837