Order Entry
Finland
ContactUsLinkComponent
Methyl 4-(3-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Methyl 4-(3-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Catalog # APOSOR33361-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-(3-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Catalog # APOSOR33361-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.719 g/mol
Storage Temperature:Ambient
CAS Number:303140-75-2
Formula:C₁₄H₁₄ClNO₃
MDL Number:MFCD00975285