5-Chloro-1-[(3-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid, tech
Catalog # APOSOR33339-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:298.121 g/mol
Storage Temperature:Ambient
CAS Number:222414-95-1
Formula:C₁₃H₉Cl₂NO₃
MDL Number:MFCD00140530