Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,4,9-Tetrahydro-1H-carbazol-1-ol
2,3,4,9-Tetrahydro-1H-carbazol-1-ol 95%
2,3,4,9-Tetrahydro-1H-carbazol-1-ol 95%
APOSOR33298-1G
:
Apollo Scientific
:
2,3,4,9-Tetrahydro-1H-carbazol-1-ol 95%
APOSOR33298-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
187.238 g/mol
:
Ambient
:
1592-62-7
:
C₁₂H₁₃NO
:
MFCD00229308