Order Entry
Finland
ContactUsLinkComponent
(2E)-1-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]-3-(dimethylamino)prop-2-en-1-one, tech
Catalog # APOSOR33030-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-1-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]-3-(dimethylamino)prop-2-en-1-one, tech
Catalog # APOSOR33030-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :268.353 g/mol
Storage Temperature:Ambient
CAS Number:692287-23-3
Formula:C₁₇H₂₀N₂O
MDL Number:MFCD03012839