Order Entry
Finland
ContactUsLinkComponent
Methyl 1-(3-chlorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylate, tech
Methyl 1-(3-chlorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylate, tech
Catalog # APOSOR33009-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 1-(3-chlorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylate, tech
Catalog # APOSOR33009-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :264.664 g/mol
Storage Temperature:Ambient
CAS Number:478063-66-0
Formula:C₁₂H₉ClN₂O₃
MDL Number:MFCD01871606