Order Entry
Finland
ContactUsLinkComponent
Ethyl 4-(4-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Ethyl 4-(4-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Catalog # APOSOR32866-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-(4-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Catalog # APOSOR32866-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :293.745 g/mol
Storage Temperature:Ambient
CAS Number:329700-15-4
Formula:C₁₅H₁₆ClNO₃
MDL Number:MFCD00664291