Order Entry
Finland
ContactUsLinkComponent
Methyl 2-methyl-5-oxo-1-(2-phenylethyl)-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Methyl 2-methyl-5-oxo-1-(2-phenylethyl)-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Catalog # APOSOR32570-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-methyl-5-oxo-1-(2-phenylethyl)-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Catalog # APOSOR32570-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.3 g/mol
Storage Temperature:Ambient
CAS Number:371144-81-9
Formula:C₁₅H₁₇NO₃
MDL Number:MFCD01893900