Order Entry
Finland
ContactUsLinkComponent
Methyl 1-[(2-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylate, tech
Methyl 1-[(2-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylate, tech
Catalog # APOSOR32390-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 1-[(2-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylate, tech
Catalog # APOSOR32390-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :277.703 g/mol
Storage Temperature:Ambient
CAS Number:320419-76-9
Formula:C₁₄H₁₂ClNO₃
MDL Number:MFCD00140817