(2S,4R)-Methyl 4-hydroxy-1-methylpyrrolidine-2-carboxylate
Catalog # APOSOR323173-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:159.183 g/mol
Storage Temperature:Ambient
CAS Number:13135-69-8
Formula:C₇H₁₃NO₃
MDL Number:MFCD18071015