1,2-Dimethyl (1R,2R)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate, tech
Catalog # APOSOR32236-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:226.269 g/mol
Storage Temperature:Ambient
CAS Number:1881275-59-7
Formula:C₁₂H₁₈O₄
MDL Number:MFCD08690904