Order Entry
Finland
ContactUsLinkComponent
1,2-Dimethyl (1R,2S)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate, tech
1,2-Dimethyl (1R,2S)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate, tech
Catalog # APOSOR32235-1G
Supplier:  Apollo Scientific
CAS Number:  
1,2-Dimethyl (1R,2S)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate, tech
Catalog # APOSOR32235-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.269 g/mol
Storage Temperature:Ambient
CAS Number:55264-91-0
Formula:C₁₂H₁₈O₄
MDL Number:MFCD02571655