Order Entry
Finland
ContactUsLinkComponent
3,5-Dimethyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid, tech
3,5-Dimethyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid, tech
Catalog # APOSOR32143-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Dimethyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid, tech
Catalog # APOSOR32143-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :230.262 g/mol
Storage Temperature:Ambient
CAS Number:108444-21-9
Formula:C₁₃H₁₄N₂O₂
MDL Number:MFCD05974918