Order Entry
Finland
ContactUsLinkComponent
Methyl 2-(4-formylphenyl)acetate 98%
Irritant
Methyl 2-(4-formylphenyl)acetate 98%
Catalog # APOSOR313093-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 2-(4-formylphenyl)acetate 98%
Catalog # APOSOR313093-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :178.185 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:96524-70-8
Formula:C₁₀H₁₀O₃
MDL Number:MFCD17018734