Order Entry
Finland
ContactUsLinkComponent
4-Amino-3-bromo-8-methoxyquinoline
4-Amino-3-bromo-8-methoxyquinoline
Catalog # APOSOR309339-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Amino-3-bromo-8-methoxyquinoline
Catalog # APOSOR309339-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :253.095 g/mol
Storage Temperature:Ambient
CAS Number:1210642-96-8
Formula:C₁₀H₉BrN₂O
MDL Number:MFCD13619788