
3-Bromo-4-chloro-7-methoxyquinoline
Catalog # APOSOR309332-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:272.526 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1203579-63-8
Formula:C₁₀H₇BrClNO
MDL Number:MFCD13192952