Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-6-chloro-8-methoxyquinoline
4-Bromo-6-chloro-8-methoxyquinoline
4-Bromo-6-chloro-8-methoxyquinoline
4-Bromo-6-chloro-8-methoxyquinoline
APOSOR309313-1G
:
Apollo Scientific
:
4-Bromo-6-chloro-8-methoxyquinoline
APOSOR309313-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
272.526 g/mol
:
Ambient
:
1189107-33-2
:
C₁₀H₇BrClNO
:
MFCD12675014