tert-Butyl 3-ethynylazetidine-1-carboxylate
Catalog # APOSOR308016-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.23 g/mol
Storage Temperature:Ambient
CAS Number:287193-01-5
Formula:C₁₀H₁₅NO₂
MDL Number:MFCD17016080