Order Entry
Finland
ContactUsLinkComponent
trans-3-(2-Hydroxypropan-2-yl)cyclobutane-1-carboxylic acid
trans-3-(2-Hydroxypropan-2-yl)cyclobutane-1-carboxylic acid
Catalog # APOSOR304385-500MG
Supplier:  Apollo Scientific
CAS Number:  
trans-3-(2-Hydroxypropan-2-yl)cyclobutane-1-carboxylic acid
Catalog # APOSOR304385-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :158.195 g/mol
Storage Temperature:Ambient
CAS Number:1940136-70-8
Formula:C₈H₁₄O₃
MDL Number:MFCD30803329