Pyridazine-3,6-diamine
Catalog # APOSOR302511-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:110.117 g/mol
Storage Temperature:Ambient
CAS Number:61070-99-3
Formula:C₄H₆N₄
MDL Number:MFCD05665950