Order Entry
Finland
ContactUsLinkComponent
4-(1,2,3-Thiadiazol-4-yl)aniline
Irritant
4-(1,2,3-Thiadiazol-4-yl)aniline
Catalog # APOSOR29027-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(1,2,3-Thiadiazol-4-yl)aniline
Catalog # APOSOR29027-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :177.226 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:121180-51-6
Formula:C₈H₇N₃S
MDL Number:MFCD00052104