Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-bromomethyl-4-cyanobenzoate
Methyl 2-bromomethyl-4-cyanobenzoate 95+%
Methyl 2-bromomethyl-4-cyanobenzoate 95+%
APOSOR2663-1G
:
Apollo Scientific
:
Methyl 2-bromomethyl-4-cyanobenzoate 95+%
APOSOR2663-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
254.08 g/mol
:
Ambient
:
165111-46-6
:
C₁₀H₈BrNO₂
:
MFCD09864855