Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-(bromomethyl)-4-cyanobenzoate
Methyl 3-(bromomethyl)-4-cyanobenzoate
Methyl 3-(bromomethyl)-4-cyanobenzoate
Irritant
Methyl 3-(bromomethyl)-4-cyanobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 3-(bromomethyl)-4-cyanobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
908562-25-4
:
Formula:
C₁₀H₈BrNO₂
MDL Number:
MFCD09839300
:
254.08 g/mol
:
GHS07
Storage Temperature:
Ambient