Order Entry
Finland
ContactUsLinkComponent
Ethyl 2-methyl-7-oxo-4,7-dihydropyrazolo[1,5-a]pyrimidine-6-carboxylate
Irritant
Ethyl 2-methyl-7-oxo-4,7-dihydropyrazolo[1,5-a]pyrimidine-6-carboxylate
Catalog # APOSOR25189-50MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 2-methyl-7-oxo-4,7-dihydropyrazolo[1,5-a]pyrimidine-6-carboxylate
Catalog # APOSOR25189-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :221.213 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:99056-35-6
Formula:C₁₀H₁₁N₃O₃
MDL Number:MFCD00067951