Order Entry
Finland
ContactUsLinkComponent
1-(4-Methyl-4,5-dihydro-1H-imidazol-2-yl)-1H-pyrazole hydroiodide
1-(4-Methyl-4,5-dihydro-1H-imidazol-2-yl)-1H-pyrazole hydroiodide
Catalog # APOSOR200146-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Methyl-4,5-dihydro-1H-imidazol-2-yl)-1H-pyrazole hydroiodide
Catalog # APOSOR200146-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :278.094 g/mol
Storage Temperature:Ambient
CAS Number:1822851-86-4
Formula:C₇H₁₀N₄·HI
MDL Number:MFCD28154661