Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2-methoxy-3-nitrobenzoic acid
5-Bromo-2-methoxy-3-nitrobenzoic acid
5-Bromo-2-methoxy-3-nitrobenzoic acid
:
Apollo Scientific
5-Bromo-2-methoxy-3-nitrobenzoic acid
:
Apollo Scientific
:
60541-89-1
:
:
C₈H₆BrNO₅
:
MFCD23701717
:
276.041 g/mol
:
176…177 °C
:
GHS07
:
Ambient