4-Chloro-N,N-dimethylaniline
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: N-(4-Chlorophenyl)dimethylamine
- CAS Number:698-69-1
- Formula:C₈H₁₀ClN
- MDL Number:MFCD07780200
- Molecular Weight:155.625 g/mol
- Boiling Pt:114…115 °C (12 mm Hg)
- Melting Pt:32…37 °C
- Density:1.049 g/cm³ (25 °C)
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: N-(4-Chlorophenyl)dimethylamine
- CAS Number:698-69-1
- Formula:C₈H₁₀ClN
- MDL Number:MFCD07780200
- Molecular Weight:155.625 g/mol
- Boiling Pt:114…115 °C (12 mm Hg)
- Melting Pt:32…37 °C
- Density:1.049 g/cm³ (25 °C)
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III



