Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Cyano-4-hydroxybenzoic acid
3-Cyano-4-hydroxybenzoic acid 95+%
3-Cyano-4-hydroxybenzoic acid 95+%
Irritant
3-Cyano-4-hydroxybenzoic acid 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Cyano-4-hydroxybenzoic acid 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
70829-28-6
:
Formula:
C₈H₅NO₃
MDL Number:
MFCD09998350
:
163.13 g/mol
:
GHS07
Storage Temperature:
Ambient