
3-Chloropyridine-4-boronic acid, neopentyl glycol ester
Catalog # APOSOR13217-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:225.48 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:915070-52-9
Formula:C₁₀H₁₃BClNO₂
MDL Number:MFCD06796314