Order Entry
Finland
ContactUsLinkComponent
Methyl 3-methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoate
Methyl 3-methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoate
Catalog # APOSOR110874-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoate
Catalog # APOSOR110874-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.29 g/mol
Storage Temperature:Ambient
CAS Number:1268618-92-3
Formula:C₁₄H₁₇NO₃
MDL Number:MFCD25976395