Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-acetyl-2H-indazole-4-carboxylate
Methyl 2-acetyl-2H-indazole-4-carboxylate
Methyl 2-acetyl-2H-indazole-4-carboxylate
APOSOR110287-1G
:
Apollo Scientific
:
Methyl 2-acetyl-2H-indazole-4-carboxylate
APOSOR110287-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
218.209 g/mol
:
GHS07
:
Ambient
:
1303890-10-9
:
C₁₁H₁₀N₂O₃
:
MFCD14635899