Methyl 2-acetyl-5,7-dibromo-2H-indazole-6-carboxylate
Catalog # APOSOR110100-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:376.001 g/mol
Storage Temperature:Ambient
CAS Number:1427460-31-8
Formula:C₁₁H₈Br₂N₂O₃
MDL Number:MFCD23159163