
Dimethyl [2-(methoxycarbonyl)ethyl]phosphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18733-15-8
:
Formula:C₆H₁₃O₅P
MDL Number:MFCD00137948
:196.138 g/mol
Boiling Pt:137…140 °C (10 mm Hg)
Density:1.21 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient