0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
t-Boc-N-Amido-PEG7-Amine
t-Boc-N-Amido-PEG7-Amine
t-Boc-N-Amido-PEG7-Amine
Irritant
t-Boc-N-Amido-PEG7-Amine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
t-Boc-N-Amido-PEG7-Amine
Supplier:
Apollo Scientific
About this item
CAS Number:
206265-98-7
:
Formula:
C₂₁H₄₄N₂O₉
MDL Number:
MFCD03453229
:
468.582 g/mol
:
GHS07
Storage Temperature:
Refrigerator