
Amino-PEG3-acid
Catalog # APOSBIPG1032-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:221.251 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:784105-33-5
Formula:C₉H₁₉NO₅
MDL Number:MFCD21607085