Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Nitro-DL-tryptophan
5-Nitro-DL-tryptophan
5-Nitro-DL-tryptophan
Irritant
5-Nitro-DL-tryptophan
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Nitro-DL-tryptophan
Supplier:
Apollo Scientific
About this item
CAS Number:
6525-46-8
:
Formula:
C₁₁H₁₁N₃O₄
MDL Number:
MFCD02683872
:
249.223 g/mol
Melting Pt:
270…272 °C
:
GHS07
Storage Temperature:
Refrigerator