Order Entry
Finland
ContactUsLinkComponent
PIG® DRI loose absorbent
PIG® DRI loose absorbent
Catalog # NEWPPLP213-1
Supplier:  NEW PIG
PIG® DRI loose absorbent
Catalog # NEWPPLP213-1
Supplier:  NEW PIG

Specifications

  • Description:
    Loose absorbent, 60% cellulose/40% kaolin
  • Absorbency:
    18,9 L
  • Pk:
    1

Specifications

About this item

Kick clay to the curb with this 100% recycled, silica-safe alternative. PIG DRI offers the same spreadability without the health risks.

  • Unlike clay, which is simply coated with liquid, fast-wicking cellulose-based kaolin granules begin to soak up liquid the moment they touch the spill
  • 100% recycled content is a byproduct of the paper-making process, ideal when green products are desired or required
  • Silica-free composition does not pose the health hazards associated with breathing in clay dust
  • Light-weight absorbent is easier than clay to carry and use without injury
  • Absorbs and retains oils, coolants, solvents and water
  • Can be incinerated after use to reduce waste or for fuels blending

Weight: 18,1 kg per bag

Volume bag: 30 L