VWR®, Centrifuge Tubes, Borosilicate Glass 3.3
Reusable centrifuge tubes made from borosilicate 3.3 glass, available with round or conical bottom, graduated or non graduated.
- Produced from borosilicate 3.3 glass tubing
- Highly resistant against chemicals and rapid temperature changes
- Autoclavable
- Withstand forces up to 3500 ×g
Graduations and inscriptions in high contrasting blue enamel. With straight rim.
: DIN 58970 (only round bottom tubes).