Malachite green oxalate solution, Sigma-Aldrich®
Catalog # 38978-100ML-F
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:927.02 g/mol
CAS Number:2437-29-8
:18015-76-4
Formula:C₅₂H₅₄N₄O₁₂
MDL Number:MFCD00011766