Order Entry
Finland
ContactUsLinkComponent
Periodic acid solution, Sigma-Aldrich®
Periodic acid solution, Sigma-Aldrich®
Catalog # 3951-100ML
CAS Number:  
Periodic acid solution, Sigma-Aldrich®
Catalog # 3951-100ML
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :227.94 g/mol
ADR:8,II
 :GHS05
Storage Temperature:Refrigerator
CAS Number:10450-60-9
Formula:H₅IO₆
MDL Number:MFCD00011344