Order Entry
Finland
ContactUsLinkComponent
 
3,3'-Dipropylthiacarbocyanine iodide ≥98%, Sigma-Aldrich®
Catalog # 318434-250MG
CAS Number:  
undefined
3,3'-Dipropylthiacarbocyanine iodide ≥98%, Sigma-Aldrich®
Catalog # 318434-250MG
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Melting Pt:288 °C (dec., lit.)
 :520.491 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:53336-12-2
Formula:C₂₃H₂₅IN₂S₂