Order Entry
Finland
ContactUsLinkComponent
Naphthol yellow S hydrate for microscopy, for the precipitation of amino acids and peptides, Sigma-Aldrich®
Naphthol yellow S hydrate for microscopy, for the precipitation of amino acids and peptides, Sigma-Aldrich®
Certificates
undefined
Naphthol yellow S hydrate for microscopy, for the precipitation of amino acids and peptides, Sigma-Aldrich®
Certificates

Specification Test Results

  • Appearance (colour):
    Light yellow to yellow and light orange to orange
  • Appearance (form):
    Powder
  • Thin layer chromatogr.:
    Corresponds to standard
  • Solubility (colour):
    Yellow to very dark yellow and orange to very dark orange
  • Solubility (turbidity):
    --
  • Solubility (method):
    0.5 g in 10 ml methanol/water 1:1
  • Wavelength (1) (UV):
    423 - 433 nm
  • Extinction (1%/1 cm) (1) (UV):
    ≥275
  • Wavelength (2) (UV):
    387 - 397 nm
  • Extinction (1%/1 cm) (2) (UV):
    ≥250
  • Solvent (UV):
    Water

Specification Test Results

Related Information