Order Entry
Finland
ContactUsLinkComponent
Rose Bengal ≥80% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Rose Bengal ≥80% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Certificates
Rose Bengal ≥80% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Certificates

Specification Test Results

  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength, C = 0.008 g/L; Water + 1 ml of 1% w/v Sodium Carbonate (Na₂CO₃):
    546 - 550 nm
  • Extinction Coefficient:
    ≥90000
  • Biological Stain Commission, Commission Certified Number::
    Pass
  • Dye Content:
    ≥80%
  • Solubility (Turbidity), c = 1 mg/ml, H₂O:
    Clear to Slightly Hazy
  • Solubility (Colour):
    Light Red to Dark Red and Pink

Specification Test Results

Related Information