Order Entry
Finland
ContactUsLinkComponent
Direct yellow 27, Sigma-Aldrich®
Direct yellow 27, Sigma-Aldrich®
Catalog # 201871-10G
CAS Number:  
Direct yellow 27, Sigma-Aldrich®
Catalog # 201871-10G
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance (Color):
    Yellow to Very Dark Yellow
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength, c=0.02g/L Water:
    224 - 230 nm
  • Extinction Coefficient:
    53000 - 57000
  • Carbon:
    Specification: Report Results
  • Nitrogen:
    Specification: Report Results
  • Sulfur:
    Specification: Report Results

Specification Test Results

About this item

 :
 :662.62 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:233-461-3
CAS Number:10190-68-8
Formula:C₂₅H₂N₄Na₂O₉S₃
MDL Number:MFCD00005778