Acid red 1, Sigma-Aldrich®
Catalog # 11640-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:509.41 g/mol
Storage Temperature:Ambient
CAS Number:3734-67-6
Formula:C₁₈H₁₅N₃O₈S₂
MDL Number:MFCD00003954